2-Ethoxy-1-naphthaleneboronic acid ≥97%

Supplier: AOBChem USA

Warning

11912-25G 11912-5G
76894-694EA 204.76 USD
76894-694 76894-696
2-Ethoxy-1-naphthaleneboronic acid ≥97%
2-Ethoxy-1-naphthaleneboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₃BO₃
MW: 216.0408 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05664211
CAS Number: 148345-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR