2,5-Difluoro-4-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

11874-1G 11874-25G 11874-500MG 11874-5G
76894-682EA 157.02 USD
76894-682 76894-684 76894-686 76894-688
2,5-Difluoro-4-methoxyphenylboronic acid ≥97%
2,5-Difluoro-4-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD04038754
CAS Number: 897958-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR