2,6-Dibromo-3-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

11701-1G 11701-25G 11701-5G 11701-500MG
76894-652EA 94.43 USD
76894-652 76894-654 76894-656 77648-384
2,6-Dibromo-3-methoxyphenylboronic acid ≥97%
2,6-Dibromo-3-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BBr₂O₃
MW: 309.75
Storage Temperature: Ambient
MDL Number: MFCD06659861
CAS Number: 850567-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR