2-hydroxy-4-methyl-5-nitrobenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

116481-1G 116481-25G 116481-500MG 116481-5G
76894-642EA 158.08 USD
76894-642 76894-644 76894-646 76894-648
2-hydroxy-4-methyl-5-nitrobenzoic acid ≥97% research grade
2-hydroxy-4-methyl-5-nitrobenzoic acid
Formula: C₈H₇NO₅
Storage Temperature: Ambient
CAS Number: 17276-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR