2,6-Dimethoxy-3-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Dimethoxypyridine-3-boronic acid, 2,6-Dimethoxypyridin-3-ylboronic acid

11538-25G 11538-5G
76894-604EA 240.83 USD
76894-604 76894-606
2,6-Dimethoxy-3-pyridineboronic acid ≥97%
2,6-Dimethoxy-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₇H₁₀BNO₄
Storage Temperature: Ambient
MDL Number: MFCD03788239
CAS Number: 221006-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR