2-Chloro-5-methyl-3-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

11437-25G 11437-5G
76894-582EA 304.48 USD
76894-582 76894-584
2-Chloro-5-methyl-3-pyridineboronic acid ≥97%
2-Chloro-5-methyl-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BClNO₂
Storage Temperature: Ambient
MDL Number: MFCD04112563
CAS Number: 913835-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR