2-Chloro-4-ethoxyphenylboronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
11410-25G
11410-5G
76894-564EA
286.45
USD
76894-564
76894-566
2-Chloro-4-ethoxyphenylboronic acid ≥97%
2-Chloro-4-ethoxyphenylboronic acid
Precaución: for Research Use Only
Formula:
C₈H₁₀BClO₃ Storage Temperature: Ambient |
MDL Number:
MFCD02684318 CAS Number: 313545-44-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...