2-Bromo-6-chloro-5-methylpyridin-3-amine ≥97% research grade

Supplier: AOBChem USA

Warning

113756-1G 113756-25G 113756-5G 113756-500MG
76894-554EA 90.18 USD
76894-554 76894-556 76894-558 77648-362
2-Bromo-6-chloro-5-methylpyridin-3-amine ≥97% research grade
2-Bromo-6-chloro-5-methylpyridin-3-amine

Precaución: for Research Use Only

Formula: C₆H₆BrClN₂
MW: 221.482 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22572520
CAS Number: 1823003-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR