2-Chloroisonicotinonitrile ≥97%

Supplier: AOBChem USA

Warning

11364-1G 11364-25G 11364-5G
76894-546EA 75.33 USD
76894-546 76894-548 76894-550
2-Chloroisonicotinonitrile ≥97%
2-Chloroisonicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₃ClN₂
Storage Temperature: Ambient
MDL Number: MFCD00174318
CAS Number: 33252-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR