2-Formyl-3-thiopheneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Formylthiophene-3-boronic acid, (2-Formylthiophen-3-yl)boronic acid

11219-25G 11219-5G
76894-532EA 418 USD
76894-532 76894-534
2-Formyl-3-thiopheneboronic acid ≥97%
2-Formyl-3-thiopheneboronic acid

Precaución: for Research Use Only

Formula: C₅H₅BO₃S
MW: 155.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01075678
CAS Number: 4347-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR