2-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-t-Butoxycarboxyphenylboronic acid, pinacol ester

10982-25G 10982-5G
76894-482EA 310.85 USD
76894-482 76894-484
2-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester ≥97%
2-(tert-Butoxycarbonyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₅BO₄
Storage Temperature: Refrigerator
MDL Number: MFCD03453053
CAS Number: 956229-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR