2-Butoxy-5-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

10895-25G 10895-5G
76894-468EA 453.01 USD
76894-468 76894-470
2-Butoxy-5-fluorophenylboronic acid ≥97%
2-Butoxy-5-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BFO₃
Storage Temperature: Ambient
MDL Number: MFCD05664232
CAS Number: 480438-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR