2-Bromo-4,5-difluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

10852-25G 10852-5G
76894-458EA 524.09 USD
76894-458 76894-460
2-Bromo-4,5-difluorophenylboronic acid ≥97%
2-Bromo-4,5-difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BBrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06411353
CAS Number: 849062-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR