2-Benzyloxy-3-bromo-5-methylphenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: (2-(Benzyloxy)-3-bromo-5-methylphenyl)boronic acid

10840-25G 10840-5G
76894-448EA 204.76 USD
76894-448 76894-450
2-Benzyloxy-3-bromo-5-methylphenylboronic acid ≥97% research grade
2-Benzyloxy-3-bromo-5-methylphenylboronic acid
Formula: C₁₄H₁₄BBrO₃
Storage Temperature: Ambient
MDL Number: MFCD06200723
CAS Number: 870777-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR