3-(2,2,2-Trifluoroacetyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

107414-1G 107414-25G 107414-500MG 107414-5G
76894-418EA 227.04 USD
76894-418 76894-420 76894-422 76894-424
3-(2,2,2-Trifluoroacetyl)benzoic acid ≥95%
3-(2,2,2-Trifluoroacetyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₅F₃O₃
MW: 218.129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13173376
CAS Number: 213598-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR