(2-Acetamidophenyl)boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Acetamidobenzeneboronic acid pinacol ester, N-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetamide, 2-Acetylaminophenylboronic acid, pinacol ester

10601-25G 10601-5G
76894-406EA 431.79 USD
76894-406 76894-408
(2-Acetamidophenyl)boronic acid pinacol ester ≥97%
(2-Acetamidophenyl)boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₀BNO₃
MW: 261.1245 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179480
CAS Number: 380430-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR