2,5-Dichloro-4-pyridineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,5-Dichloropyridine-4-boronic acid pinacol ester, 2,5-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

10543-1G 10543-25G 10543-500MG 10543-5G
76894-396EA 129.43 USD
76894-396 76894-398 76894-400 76894-402
2,5-Dichloro-4-pyridineboronic acid pinacol ester ≥97%
2,5-Dichloro-4-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₄BCl₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD06798256
CAS Number: 1073353-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR