2-Fluoro-5-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-5-formylbenzeneboronic acid

10527-25G 10527-5G
76894-388EA 183.54 USD
76894-388 76894-390
2-Fluoro-5-formylphenylboronic acid ≥97%
2-Fluoro-5-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BFO₃
MW: 167.93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03095100
CAS Number: 352534-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR