2-Fluoroisonicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoropyridine-4-carboxaldehyde

10485-1G 10485-25G 10485-5G
76894-348EA 62.6 USD
76894-348 76894-350 76894-352
2-Fluoroisonicotinaldehyde ≥97%
2-Fluoroisonicotinaldehyde
Formula: C₆H₄FNO
MW: 125.1000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06410677
CAS Number: 131747-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR