2-Fluoro-6-(trifluoromethyl)nicotinaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

102253-1G 102253-500MG 102253-5G
76894-316EA 760.67 USD
76894-316 76894-318 76894-320
2-Fluoro-6-(trifluoromethyl)nicotinaldehyde ≥97% research grade
2-Fluoro-6-(trifluoromethyl)nicotinaldehyde
Formula: C₇H₃F₄NO
MW: 193.098 g/mol
Storage Temperature: Refrigerator
CAS Number: 1227599-54-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR