2-Chloro-6-isopropylpyridine ≥97%

Supplier: AOBChem USA

Warning

10194-25G 10194-5G
76894-284EA 595.17 USD
76894-284 76894-286
2-Chloro-6-isopropylpyridine ≥97%
2-Chloro-6-isopropylpyridine

Precaución: for Research Use Only

Formula: C₈H₁₀ClN
Storage Temperature: Ambient
MDL Number: MFCD08689614
CAS Number: 120145-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR