2-Chloro-5-cyanophenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

10064-25G 10064-5G
76894-218EA 403.15 USD
76894-218 76894-220
2-Chloro-5-cyanophenylboronic acid ≥97% research grade
2-Chloro-5-cyanophenylboronic acid
Formula: C₇H₅BClNO₂
Storage Temperature: Ambient
MDL Number: MFCD06656271
CAS Number: 936249-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR