2-Chloro-3-methyl-5-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

10061-25G 10061-5G
76894-212EA 209 USD
76894-212 76894-214
2-Chloro-3-methyl-5-pyridineboronic acid ≥97%
2-Chloro-3-methyl-5-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BClNO₂
Storage Temperature: Ambient
MDL Number: MFCD03411564
CAS Number: 1003043-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR