2-Bromo-4-hydroxypyridine ≥97%

Supplier: AOBChem USA

Warning

100483-25G 100483-5G
76894-200EA 460.44 USD
76894-200 76894-202
2-Bromo-4-hydroxypyridine ≥97%
2-Bromo-4-hydroxypyridine
Formula: C₅H₄BrNO
Storage Temperature: Ambient
MDL Number: MFCD00234040
CAS Number: 36953-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR