2,6-Dichloro-3-pyridineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Dichloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2,6-Dichloropyridine-3-boronic acid pinacol ester

10039-1G 10039-25G 10039-500MG 10039-5G
76894-184EA 148.53 USD
76894-184 76894-186 76894-188 76894-190
2,6-Dichloro-3-pyridineboronic acid pinacol ester ≥97%
2,6-Dichloro-3-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₄BCl₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD06798261
CAS Number: 1073371-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR