4-(Cyclopropylmethoxy)-1-fluoro-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

95353-25G 95353-5G
76918-218EA 368.14 USD
76918-218 76918-220
4-(Cyclopropylmethoxy)-1-fluoro-2-methoxybenzene ≥97%
4-(Cyclopropylmethoxy)-1-fluoro-2-methoxybenzene
Formula: C₁₁H₁₃FO₂
MW: 196.218 g/mol
Storage Temperature: Ambient
CAS Number: 1360807-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR