3-Bromo-2-Chloro-6-(Methylsulfonyl)Pyridine ≥97%

Supplier: AOBChem USA

Warning

91266-1G 91266-25G 91266-500MG 91266-5G
76918-204EA 354.35 USD
76918-204 76918-206 76918-208 76918-210
3-Bromo-2-Chloro-6-(Methylsulfonyl)Pyridine ≥97%
3-Bromo-2-Chloro-6-(Methylsulfonyl)Pyridine
Formula: C₆H₅BrClNO₂S
MW: 270.531 g/mol
Storage Temperature: Ambient
CAS Number: 2918828-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR