4-(1,1-Difluoroethyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

90567-1G 90567-25G 90567-500MG 90567-5G
76918-188EA 256.74 USD
76918-188 76918-190 76918-192 76918-194
4-(1,1-Difluoroethyl)benzoic acid ≥95%
4-(1,1-Difluoroethyl)benzoic acid
Formula: C₉H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD20643344
CAS Number: 55805-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR