4-Bromo-2-chloro-1-(cyclopropylmethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

89932-25G 89932-5G
76918-176EA 605.78 USD
76918-176 76918-178
4-Bromo-2-chloro-1-(cyclopropylmethoxy)benzene ≥97%
4-Bromo-2-chloro-1-(cyclopropylmethoxy)benzene
Formula: C₁₀H₁₀BrClO
MW: 261.543 g/mol
Storage Temperature: Ambient
CAS Number: 1369869-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR