3-(9H-carbazol-9-yl)-5-chlorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

88291-1G 88291-500MG 88291-5G 88291-25G
76918-130EA 1631.67 USD
76918-130 76918-132 77630-806 77660-480
3-(9H-carbazol-9-yl)-5-chlorobenzaldehyde ≥97%
3-(9H-carbazol-9-yl)-5-chlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₉H₁₂ClNO
Storage Temperature: Refrigerator
MDL Number: MFCD34817777
CAS Number: 2921724-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR