3,5-Dibromo-4-ethoxyaniline ≥97% research grade

Supplier: AOBChem USA

Warning

88002-1G 88002-25G 88002-500MG 88002-5G
76918-116EA 439.22 USD
76918-116 76918-118 76918-120 76918-122
3,5-Dibromo-4-ethoxyaniline ≥97% research grade
3,5-Dibromo-4-ethoxyaniline
Formula: C₈H₉Br₂NO
MW: 294.971 g/mol
Storage Temperature: Ambient
CAS Number: 52765-17-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR