3-Acetyl-2,6-dichlorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

87612-1G 87612-25G 87612-500MG 87612-5G
76918-104EA 215.37 USD
76918-104 76918-106 76918-108 76918-110
3-Acetyl-2,6-dichlorobenzaldehyde ≥97%
3-Acetyl-2,6-dichlorobenzaldehyde
Formula: C₉H₆Cl₂O₂
Storage Temperature: Ambient
CAS Number: 2383945-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR