4-Bromo-2,5-dichlorobenzonitrile ≥97%

Supplier: AOBChem USA

Warning

85377-1G 85377-500MG 85377-5G
76918-078EA 512.42 USD
76918-078 76918-080 76918-082
4-Bromo-2,5-dichlorobenzonitrile ≥97%
4-Bromo-2,5-dichlorobenzonitrile
Formula: C₇H₂BrCl₂N
Storage Temperature: Ambient
CAS Number: 1349717-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR