3-Bromo-2,5-difluorophenol ≥97%

Supplier: AOBChem USA

Warning

82288-1G 82288-500MG 82288-5G
76918-058EA 530.45 USD
76918-058 76918-060 76918-062
3-Bromo-2,5-difluorophenol ≥97%
3-Bromo-2,5-difluorophenol
Formula: C₆H₃BrF₂O
MW: 208.988 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28528419
CAS Number: 1783357-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR