3-Chloro-5-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

80995-1G 80995-25G 80995-500MG 80995-5G
76918-042EA 217.49 USD
76918-042 76918-044 76918-046 76918-048
3-Chloro-5-iodopyridine ≥97%
3-Chloro-5-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₃ClIN
MW: 239.4415 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08235133
CAS Number: 77332-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR