3,5-Difluoro-N-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

80463-1G 80463-25G 80463-5G
76918-028EA 70.02 USD
76918-028 76918-030 76918-032
3,5-Difluoro-N-methoxy-N-methylbenzamide ≥97%
3,5-Difluoro-N-methoxy-N-methylbenzamide
Formula: C₉H₉F₂NO₂
Storage Temperature: Ambient
CAS Number: 874889-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR