3-Bromo-5-cyanobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

75981-1G 75981-25G 75981-500MG 75981-5G
76917-984EA 158.08 USD
76917-984 76917-986 76917-988 76917-990
3-Bromo-5-cyanobenzoic acid ≥95%
3-Bromo-5-cyanobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄BrNO₂
MW: 226.027 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09261037
CAS Number: 453566-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR