4-(Difluoromethyl)-3-fluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

75093-1G 75093-25G 75093-500MG 75093-5G
76917-968EA 441.34 USD
76917-968 76917-970 76917-972 76917-974
4-(Difluoromethyl)-3-fluorobenzoic acid ≥95%
4-(Difluoromethyl)-3-fluorobenzoic acid
Formula: C₈H₅F₃O₂
MW: 190.1193 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28522914
CAS Number: 1780188-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR