3'-Formylbiphenyl-4-carboxylic acid 95%

Supplier: AOBChem USA

Warning

Synonyms: 3'-Formyl-[1,1'-biphenyl]-4-carboxylic acid

72525-1G 72525-25G 72525-5G 72525-500MG
76917-908EA 82.76 USD
76917-908 76917-910 76917-912 77651-974
3'-Formylbiphenyl-4-carboxylic acid 95%
3'-Formylbiphenyl-4-carboxylic acid
Formula: C₁₄H₁₀O₃
MW: 226.2300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039069
CAS Number: 222180-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR