3-Chloro-2-fluoro-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

69824-1G 69824-25G 69824-500MG 69824-5G
76917-886EA 352.22 USD
76917-886 76917-888 76917-890 76917-892
3-Chloro-2-fluoro-4-methylbenzaldehyde ≥97%
3-Chloro-2-fluoro-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆ClFO
Storage Temperature: Refrigerator
MDL Number: MFCD28751754
CAS Number: 1383951-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR