4-Amino-3-chloro-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

69305-1G 69305-25G 69305-5G
76917-874EA 66.84 USD
76917-874 76917-876 76917-878
4-Amino-3-chloro-5-methylbenzoic acid ≥97%
4-Amino-3-chloro-5-methylbenzoic acid
Formula: C₈H₈ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD01313709
CAS Number: 157069-52-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR