4-Bromo-1-(4-methylphenyl)-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

69114-25G 69114-5G
76917-862EA 360.71 USD
76917-862 76917-864
4-Bromo-1-(4-methylphenyl)-1H-pyrazole ≥97%
4-Bromo-1-(4-methylphenyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂
Storage Temperature: Ambient
MDL Number: MFCD09475854
CAS Number: 957034-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR