3-Bromo-2-iodothiophene ≥97%

Supplier: AOBChem USA

Warning

68078-1G 68078-25G 68078-500MG 68078-5G
76917-834EA 186.72 USD
76917-834 76917-836 76917-838 76917-840
3-Bromo-2-iodothiophene ≥97%
3-Bromo-2-iodothiophene

Precaución: for Research Use Only

Formula: C₄H₂BrIS
MW: 288.932 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11848091
CAS Number: 60404-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR