4-(3-Nitrophenyl)morpholine ≥97%

Supplier: AOBChem USA

Warning

67882-25G 67882-5G
76917-822EA 336.31 USD
76917-822 76917-824
4-(3-Nitrophenyl)morpholine ≥97%
4-(3-Nitrophenyl)morpholine
Formula: C₁₀H₁₂N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD03195994
CAS Number: 116922-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR