3-Formyl-4-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

66339-1G 66339-500MG 66339-5G
76917-796EA 525.15 USD
76917-796 76917-798 76917-800
3-Formyl-4-isopropoxybenzoic acid ≥97%
3-Formyl-4-isopropoxybenzoic acid
Formula: C₁₁H₁₂O₄
MW: 208.211 g/mol
Storage Temperature: Refrigerator
CAS Number: 1289061-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR