5-Bromo-2-chlorobenzal fluoride ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-1-chloro-2-(difluoromethyl)benzene

66264-1G 66264-500MG 66264-5G
76917-782EA 530.45 USD
76917-782 76917-784 76917-786
5-Bromo-2-chlorobenzal fluoride ≥97%
5-Bromo-2-chlorobenzal fluoride
Formula: C₇H₄BrClF₂
MW: 241.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698636
CAS Number: 627527-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR