3-Bromo-2,5-dimethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

64867-1G 64867-25G 64867-500MG 64867-5G
76917-714EA 228.1 USD
76917-714 76917-716 76917-718 76917-720
3-Bromo-2,5-dimethoxybenzaldehyde ≥97%
3-Bromo-2,5-dimethoxybenzaldehyde
Formula: C₉H₉BrO₃
MW: 245.0699 g/mol
Storage Temperature: Ambient
CAS Number: 68216-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR