4-(Benzyloxy)-2-bromo-1-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

63781-1G 63781-25G 63781-500MG 63781-5G
76917-698EA 157.02 USD
76917-698 76917-700 76917-702 76917-704
4-(Benzyloxy)-2-bromo-1-methoxybenzene ≥97%
4-(Benzyloxy)-2-bromo-1-methoxybenzene
Formula: C₁₄H₁₃BrO₂
MW: 293.1558 g/mol
Storage Temperature: Ambient
CAS Number: 79352-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR