3-Chloroquinolin-7-amine ≥97%

Supplier: AOBChem USA

Warning

63002-1G 63002-500MG 63002-5G
76917-676EA 593.05 USD
76917-676 76917-678 76917-680
3-Chloroquinolin-7-amine ≥97%
3-Chloroquinolin-7-amine
Formula: C₉H₇ClN₂
Storage Temperature: Ambient
MDL Number: MFCD20259285
CAS Number: 1354222-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR