3-Formyl-5-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

62829-1G 62829-25G 62829-500MG 62829-5G
76917-660EA 264.17 USD
76917-660 76917-662 76917-664 76917-666
3-Formyl-5-methoxybenzoic acid ≥97%
3-Formyl-5-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈O₄
Storage Temperature: Ambient
MDL Number: MFCD11042979
CAS Number: 367519-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR